Order Entry
Belgium
ContactUsLinkComponent
 
Analytical HPLC columns, µBondapak® Phenyl
  WATEWAT027198
 :  WATERS
Analytical HPLC columns, µBondapak® Phenyl
  WATEWAT027198
 :  WATERS

 

  • Column Format:
    Analytical
  • Ø int.:
    3,90 mm
  • Particle Size:
    10 µm
  • Pore size:
    125 Å
  • Length:
    300 mm
  • Phase:
    Phenyl
  • Pk:
    1