Gel permeation chromatography columns, Styragel® HR
Supplier: WATERS
Certificates
About this item
Styragel® HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterising samples in the low to mid molecular range where high resolution is critical.
Specifications
- Catalog No:
- WATEWAT044222
- WATEWAT044228
- WATEWAT044237
- WATEWAT044229
- WATEWAT054405
- WATEWAT044231
- WATEWAT054415
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12