Order Entry
Belgium
ContactUsLinkComponent
Gel permeation chromatography clean-up columns, Envirogel™
Gel permeation chromatography clean-up columns, Envirogel™
Catalog # WATEWAT036555
Supplier:  WATERS
Gel permeation chromatography clean-up columns, Envirogel™
Catalog # WATEWAT036555
Supplier:  WATERS

Specifications

  • Column Format:
    Preparative
  • Ø int.:
    19,00 mm
  • Particle Size:
    15 µm
  • Description:
    Envirogel™ GPC
  • Length:
    150 mm
  • Seq:
    01
  • pH range:
    2 - 12
  • Exclusion limit:
    100 - 10 000
  • Pk:
    1

Specifications

About this item

The Envirogel™ High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences such as lipids and natural resins from environmental samples as specified in EPA method 3640A.