Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Technical pH buffer solutions, HI 5000 series

 :   Hanna
Technical pH buffer solutions, HI 5000 series
 :  Hanna
 
Technical pH buffer solutions, HI 5000 series

 

 
  • Colour coded labels
  • ±0,01 pH

 : Technical buffer solutions with certificate.

 

Product Family Options

 

  • pH value
    pH accuracy
    Reference temperature
    Pk
     
     
  • 1,00
    ±0,01
    25 °C
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - 43,30 €
    43,30 €
    1 * 500 mL
       43,30 €

  • 8,00
    ±0,01
    25 °C
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - 43,30 €
    43,30 €
    1 * 500 mL
       43,30 €

  • 10,01
    ±0,01
    25 °C
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - 59,50 €
    59,50 €
    1 * 500 mL
       59,50 €