Transport boxes, SYSTAINER T-Loc
Catalog # 216-1622
Supplier: TRANSPOSAFE
Specifications
- W×D×H:400×300×210 mm
- Pk:1
Specifications
About this item
Durable ABS, light grey.
- The case is robust
- Available in four sizes
- The cases can be attached to each other easily, using the quick-acting closure
All Systainer T-Loc are sturdy and impact-resistant as well as dust and splashwater-proof. Every component of the Systainer T-Loc such as body, lid, handle or catches can be replaced easily and thus guarantees a long product life.