Cold traps, long version, type KFL
About this item
These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
- Available with or without spouts
- Complete versions include Dewar flask and plastic ring
The condensation area of the cold traps has been extended in order to trap more condensation. In addition to this, the complete cold trap versions also have a larger plastic ring and a bigger Dewar flask.
An upgraded version is a long version cold trap with an outlet spout. These cold traps are constructed in the same way as standard cold traps but have a GL 32 glass thread together with a screw-on cap, which is the actual spout. With the help of this spout the condensate can easily be poured out of the cold trap. Therefore the cold trap is easily to clean.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585 (DURAN made by Schott).
4 Options Available Below
Product Details & Documents
These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
- Available with or without spouts
- Complete versions include Dewar flask and plastic ring
The condensation area of the cold traps has been extended in order to trap more condensation. In addition to this, the complete cold trap versions also have a larger plastic ring and a bigger Dewar flask.
An upgraded version is a long version cold trap with an outlet spout. These cold traps are constructed in the same way as standard cold traps but have a GL 32 glass thread together with a screw-on cap, which is the actual spout. With the help of this spout the condensate can easily be poured out of the cold trap. Therefore the cold trap is easily to clean.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585 (DURAN made by Schott).
Product Family Options
Product Information
- AvailabilityPrice
- KGWA17362Cold trap, KFL 29-GL, complete with Dewar and plastic ring, without spout, borosilicate glass 3.3, with threaded glass joint GL 18 and PTFE olive, condensate cap. 250 ml and coolant cap. 2000 mlCold trap, KFL 29-GL, complete with Dewar and plastic ring, without spout, borosilicate glass 3.3, with threaded glass joint GL 18 and PTFE olive, condensate cap. 250 ml and coolant cap. 2000 mlLog in for availability1 * 1 ST650,00 €
- KGWA17380Cold trap, KFL 29-K-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml and coolant cap. 2000 mlCold trap, KFL 29-K-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml and coolant cap. 2000 mlLog in for availability1 * 1 ST723,00 €
- KGWA17381Cold trap, KFL 29-OK-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 mlCold trap, KFL 29-OK-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 mlLog in for availability1 * 1 ST753,00 €
- KGWA17382Cold trap, KFL 29-GL-A, with spout, complete with Dewar and plastic ring, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml and coolant cap. 2000 mlCold trap, KFL 29-GL-A, with spout, complete with Dewar and plastic ring, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml and coolant cap. 2000 mlLog in for availability1 * 1 ST713,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cold trap, KFL SL 29-GL, spare part, without spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, condensate cap. 250 mlCold trap, KFL SL 29-GL, spare part, without spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, condensate cap. 250 mlKGWA17367Log in for availability1 * 1 ST - 283,00 €
Product Information
- Catalog #AvailabilityPrice
- Cold trap SL 29-O-A, spare part, with spout, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 mlCold trap SL 29-O-A, spare part, with spout, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 mlKGWA17386Log in for availability1 * 1 ST - 386,00 €
Product Information
- Catalog #AvailabilityPrice
- Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 mlCold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 mlKGWA17387Log in for availability1 * 1 ST - 347,00 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.