Orders LinkContactUsLinkComponent
Prefilter attachment
Prefilter attachment
Prefilter attachment, Stainless steel
Catalog # SART16807
Supplier:   Sartorius
Prefilter attachment
Catalog # SART16807
Supplier:  Sartorius
Prefilter attachment
Prefilter attachment
Prefilter attachment, Stainless steel
Price
276,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Allows the separation of coarse solids from samples with bacteriological germ detection in a single operation, before and during the actual germ-retaining filtration step.
Specifications
  • Description:
    Prefilter attachment
  • Material:
    Stainless steel
  • Seq:
    01