Orders LinkContactUsLinkComponent
Sampling spatula, Sterileware®, SP Bel-Art
Sampling spatula, Sterileware®, SP Bel-Art
Sampling spatula, Sterileware®, SP Bel-Art
Certificates
Sampling spatula, Sterileware®, SP Bel-Art
Sampling spatula, Sterileware®, SP Bel-Art
2 Options Available Below
Product Details & Documents
One-piece spatulas, moulded from FDA grade, high impact PS.
  • Sterilised by gamma irradiation, lot stamped for accurate lot tracing, shipped with a Certificate of Processing for Sterilisation
V-shaped spatula for sampling dislodge caked or crystallised reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄"). The sturdy point is perfect for taking sterile samples during process quality inspections, and for piercing plastic-lined paper bags or similar packages.
Product Family Options
Product Information
  • Sterile
    Length
    Blade length
    Capacity
    Colour
    Availability
    Price
  • +
    229 mm
    153 mm
    15 ml
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 338,00 €
    338,00 €
    1 * 100 ST
       338,00 €
  • +
    356 mm
    254 mm
    23 ml
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 245,00 €
    245,00 €
    1 * 50 ST
       245,00 €