Orders LinkContactUsLinkComponent
Utility carts
Utility carts
Supplier:   Palbam
Utility carts
Supplier:  Palbam
Certificates
Utility carts
10 Options Available Below
Product Details & Documents
These cleanroom carts are designed to withstand heavy loads.
  • Up to 60 kg per shelf
  • Integral raised ledges prevent parts from rolling off and are easy to wipe down
  • Choice of two or three shelves
  • Solid or perforated
  • White nylon 130 mm (5") cleanroom castors (Two with brakes) supplied with every cart
Stainless steel 304 with electropolished finish. Special non-marking, smooth running.
Delivery information: Typically available from stock for direct shipment from the supplier. Shipped flat-packed with assembly instructions.
Product Family Options
Cleanroom carts with solid shelves
Product Information
  • W×D×H
    Shelves
    Availability
    Price
  • 139-0588
    Cleanroom utility cart
    Cleanroom utility cart
    550×550×990 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 920,00 €
    1 920,00 €
    1 * 1 ST
       1 920,00 €
  • 139-0589
    Cleanroom utility cart
    Cleanroom utility cart
    865×550×990 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 220,00 €
    2 220,00 €
    1 * 1 ST
       2 220,00 €
  • PALBUC-85-2-ESD
    Cleanroom utility cart with casters
    Cleanroom utility cart with casters
    865×550×990 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €
  • 139-0793
    Cleanroom utility cart
    Cleanroom utility cart
    550×550×990 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 139-0590
    Cleanroom utility cart
    Cleanroom utility cart
    865×550×990 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Cleanroom carts with perforated shelves
Product Information
  • W×D×H
    Shelves
    Availability
    Price
  • PALBUCP-55-2
    Cleanroom utility cart
    Cleanroom utility cart
    550×550×990 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 139-0591
    Cleanroom utility cart
    Cleanroom utility cart
    865×550×990 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 200,00 €
    2 200,00 €
    1 * 1 ST
       2 200,00 €
  • 139-0794
    Cleanroom utility cart
    Cleanroom utility cart
    550×550×990 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 139-0795
    Cleanroom utility cart
    Cleanroom utility cart
    865×550×990 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • PALBUCP-55-3-ESD
    Cleanroom utility cart
    Cleanroom utility cart
    550×550×990 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 370,00 €
    2 370,00 €
    1 * 1 ST
       2 370,00 €