Orders LinkContactUsLinkComponent
Single channel pipettes, mechanical, variable volume, VITLAB®
Single channel pipettes, mechanical, variable volume, VITLAB®
Supplier:   VITLAB
Single channel pipettes, mechanical, variable volume, VITLAB®
Supplier:  VITLAB
Certificates
Single channel pipettes, mechanical, variable volume, VITLAB®
Single channel pipettes, mechanical, variable volume, VITLAB®
7 Options Available Below
Product Details & Documents
These piston-operated manual pipettes are ideal for demanding laboratory applications.
  • Central, large pipetting button for easy, uniform piston movement
  • Large, ergonomic eject button for rapid replacement of the tips
  • Simple volume adjustment for right- and left-handers
  • Precise, 4-position volume indicator, with integrated zoom function for optimal readability
  • Colour-coded for easy selection of the right pipette tip
  • Ergonomic finger loop for convenient handling
  • Integrated calibration function for easy calibration without the need for tools
  • Corrosion-resistant pistons and ejectors for long product life
  • Completely autoclavable at 121 °C
DE-M marked
Delivery information: Supplied with silicone oil resp. grease (Size 5 and 10 ml), sample bag with pipette tips, quality certificate, and instruction manual
Product Family Options
Product Information
  • Volume
    Accuracy
    Imprecision
    Recommended tips
    Availability
    Price
  • 0,5 - 10 µl
    ±1,0 %
    ≤0,5 %
    20 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 468,00 €
    468,00 €
    1 * 1 ST
       468,00 €
  • 2 - 20 µl
    ±0,8 %
    ≤0,4 %
    200 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 418,00 €
    418,00 €
    1 * 1 ST
       418,00 €
  • 10 - 100 µl
    ±0,6 %
    ≤0,2 %
    200/300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 418,00 €
    418,00 €
    1 * 1 ST
       418,00 €
  • 20 - 200 µl
    ±0,6 %
    ≤0,2 %
    200/300 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 418,00 €
    418,00 €
    1 * 1 ST
       418,00 €
  • 100 - 1000 µl
    ±0,6 %
    ≤0,2 %
    1000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 418,00 €
    418,00 €
    1 * 1 ST
       418,00 €
  • 500 - 5000 µl
    ±0,6 %
    ≤0,2 %
    5000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 468,00 €
    468,00 €
    1 * 1 ST
       468,00 €
  • 1000 - 10000 µl
    ±0,6 %
    ≤0,2 %
    10000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 468,00 €
    468,00 €
    1 * 1 ST
       468,00 €