Orders LinkContactUsLinkComponent
Sample vials, closed-top
Sample vials, closed-top
   Sustainable Options Available
Supplier:   EP SCIENTIFIC
Sample vials, closed-top
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
Sample vials, closed-top
9 Options Available Below
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With closed-top white PP cap bonded with PTFE-lined silicone septum for secure sealing
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CoA is not included.
Specifications for Product Family
Specifications
  • Catalog No:
  • EPSCC126-0020
  • EPSCC136-0040
  • EPSCC226-0020
  • EPSCC236-0040
  • EPSCC326-0020
  • EPSCC336-0040
  • EPSCC146-0040
  • EPSCC246-0040
  • EPSCC346-0040
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Clear
Product Information
  • Capacity
    Colour
    Cap included
    Cap type
    Cap Size
    Septum
    Availability
    Price
  • 20,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 157,00 €
    157,00 €
    1 * 72 ST
       157,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC126-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 40,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 235,00 €
    235,00 €
    1 * 72 ST
       235,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC136-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 20,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 294,00 €
    294,00 €
    1 * 72 ST
       294,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC226-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 40,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 322,00 €
    322,00 €
    1 * 72 ST
       322,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC236-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 20,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 282,00 €
    282,00 €
    1 * 72 ST
       282,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC326-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 40,00 ml
    Clear
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 342,00 €
    342,00 €
    1 * 72 ST
       342,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC336-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Amber
Product Information
  • Capacity
    Colour
    Cap included
    Cap type
    Cap Size
    Septum
    Availability
    Price
  • 40,00 ml
    Amber
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 200,00 €
    200,00 €
    1 * 72 ST
       200,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC146-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 40,00 ml
    Amber
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 294,00 €
    294,00 €
    1 * 72 ST
       294,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC246-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 40,00 ml
    Amber
    Yes
    Closed
    24-414
    PTFE-faced
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 342,00 €
    342,00 €
    1 * 72 ST
       342,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC346-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact