ContactUsLinkComponent
Cleanroom sleeve protectors, Kimtech™ A5
Cleanroom sleeve protectors, Kimtech™ A5
Catalog # 113-7733
Supplier:  KIMBERLY CLARK
   Waste Reducing
undefined
Cleanroom sleeve protectors, Kimtech™ A5
Catalog # 113-7733
Supplier:  KIMBERLY CLARK
   Waste Reducing
Specifications
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Length:
    457 mm
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    100 PAIR
  • Cleanroom class:
    100/ISO 5
Specifications
About this item
These sterile sleeve protectors are made of durable SMS fabric which provides a cloth-like feel. A highly visible blue line along the inside of the sleeve signals the proper place to grasp while donning, helping workers avoid touching the outside.
  • Thumb loop keeps sleeve from rolling back and exposing arm or wrist
  • Tunnelled elastic and bound seams
  • Triple-bagged
Suitable for Class 100, ISO Class 5 or higher cleanrooms.
Certifications: PPE Cat I (EU) 2016/425.