Capillary columns, traditional phases, SPB-5, Supelco®
: Merck
These bonded poly(5% diphenyl/95% dimethylsiloxane) low phenyl content columns improve thermal stability of the phase, while still providing essentially a boiling point elution order, and a slight increase in selectivity, especially for aromatic compounds.
- Temperature limit from –60 to 320 °C
: Meets USP G27 and G36 requirements.
Frequently Bought Together