Cleanroom Basic Chairs
About this item
In order to create 'clean' or 'sterile' rooms, measures have to be implemented to prevent products and processes from becoming contaminated. This is a key requirement of the microelectronics sector, the pharmaceuticals industry, microsystem production, optics, medical engineering and healthcare. That is why bimos has worked together with industry and research specialists to develop its innovative range of cleanroom chairs for use in extreme conditions.
- The smooth, sealed surfaces of the seat and backrest shells prevent swirl even when there is a constant flow of air through the filter system and stop the build-up of particles on the chair
- The synthetic leather, upholstery and upholstery support are permanently bonded using special foam technology to ensure that no particles can be emitted
- Supplied with a brilliantly polished and abrasion resistant aluminium five star base
- The base frame, which is made from die-cast aluminium, is brilliantly polished with conductive castors or glides for hard floors
- The seat support and backrest cover are made of plastic
- Available in three versions: With contact backrest, contact backrest with tilting seat, and asynchronous mechanism with weight regulation
7 Options Available Below
Product Details & Documents
In order to create 'clean' or 'sterile' rooms, measures have to be implemented to prevent products and processes from becoming contaminated. This is a key requirement of the microelectronics sector, the pharmaceuticals industry, microsystem production, optics, medical engineering and healthcare. That is why bimos has worked together with industry and research specialists to develop its innovative range of cleanroom chairs for use in extreme conditions.
- The smooth, sealed surfaces of the seat and backrest shells prevent swirl even when there is a constant flow of air through the filter system and stop the build-up of particles on the chair
- The synthetic leather, upholstery and upholstery support are permanently bonded using special foam technology to ensure that no particles can be emitted
- Supplied with a brilliantly polished and abrasion resistant aluminium five star base
- The base frame, which is made from die-cast aluminium, is brilliantly polished with conductive castors or glides for hard floors
- The seat support and backrest cover are made of plastic
- Available in three versions: With contact backrest, contact backrest with tilting seat, and asynchronous mechanism with weight regulation
Product Family Options
Cleanroom Basic 2 with castors
Product Information
- HeightFoot styleFoot ringAvailabilityPrice
- BIMO9140-2571-807Cleanroom Basic 2 chair with stop and go castors, in skai® ESD synthetic leather, black, with contact backrest, backrest height: 430 mmCleanroom Basic 2 chair with stop and go castors, in skai® ESD synthetic leather, black, with contact backrest, backrest height: 430 mm470 - 610 mmCastorsLog in for availability1 * 1 ST573,00 €
- 139-0580Cleanroom Basic 2 chair, in skai® ESD synthetic leather, black, with contact backrest with tilting seat, backrest height: 530 mmCleanroom Basic 2 chair, in skai® ESD synthetic leather, black, with contact backrest with tilting seat, backrest height: 530 mm470 - 610 mmCastorsLog in for availability1 * 1 ST646,00 €
- 139-0582Cleanroom Basic 2 chair, in skai® ESD synthetic leather, black, with synchronous mechanism with weight regulation, backrest height: 530 mmCleanroom Basic 2 chair, in skai® ESD synthetic leather, black, with synchronous mechanism with weight regulation, backrest height: 530 mm490 - 630 mmCastorsLog in for availability1 * 1 ST688,00 €
Cleanroom Basic 3 with glides
Product Information
- HeightFoot styleFoot ringAvailabilityPrice
- BIMO9140-2571-812Cleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest, backrest height: 430 mmCleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest, backrest height: 430 mm470 - 610 mmGlidesLog in for availability1 * 1 ST548,00 €
- 139-0581Cleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest with tilting seat, backrest height: 530 mmCleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest with tilting seat, backrest height: 530 mm630 - 890 mmGlides+Log in for availability1 * 1 ST770,00 €
- 139-0583Cleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with synchronous mechanism with weight regulation, backrest height: 530 mmCleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with synchronous mechanism with weight regulation, backrest height: 530 mm660 - 910 mmGlides+Log in for availability1 * 1 ST812,00 €
- BIMO91416902525807Cleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest, adjustable backrest heightCleanroom Basic 3 chair, in skai® ESD synthetic leather, black, with contact backrest, adjustable backrest height620 - 890 mmGlides+Log in for availability1 * 1 ST710,00 €