Order Entry
Belgium
ContactUsLinkComponent
SMD tweezers, Erem®
SMD tweezers, Erem®
Catalog # BSNC40SA
Supplier:  Basan
undefined
SMD tweezers, Erem®
Catalog # BSNC40SA
Supplier:  Basan

Specifications

  • Type:
    Straight, round tips (Ø 0,4 mm)
  • Length:
    110 mm
  • Pk:
    1

Specifications

About this item

Stainless steel. These tweezers are ideal for the gripping and holding of round components and wires.

  • Non rusting, heat resistant, and anti-acid proof surface
  • Anti-magnetic
  • With blunt tips to prevent damage to printed-circuit boards
  • Serrated finger grips for a better grip