ContactUsLinkComponent
Ultrasonic processors for small scale samples, Q55 and Q125
Ultrasonic processors for small scale samples, Q55 and Q125
Fournisseur:  QSONICA LLC
Certificats
undefined
Ultrasonic processors for small scale samples, Q55 and Q125
Fournisseur:  QSONICA LLC
Spécifications
  • Catalog No:
  • 432-0236
  • 432-0235
  • 432-0232
  • 432-0231
  • Model:
  • Q55
  • Q55
  • Q125
  • Q125
  • Frequency:
  • 20 kHz
  • 20 kHz
  • 20 kHz
  • 20 kHz
  • Max. output power:
  • 55 W
  • 55 W
  • 125 W
  • 125 W
  • Timer:
  • -
  • -
  • Up to 10 hours
  • Up to 10 hours
  • Programmable:
  • -
  • -
  • Time and amplitude
  • Time and amplitude
  • W×D×H:
  • 203×190×146 mm
  • 203×190×146 mm
  • 203×350×146 mm
  • 203×350×146 mm
  • Interval time:
  • -
  • -
  • 1 second to 1 minute
  • 1 second to 1 minute
Spécifications
Informations associées
Probes and horns
Informations sur le produit
Frequently Bought Together