Flasks, round bottom, narrow neck, beaded rim
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass 3,3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.
Frequently Bought Together