ESD safety shoes, clogs, X-Light, 7131041/7131042
Supplier: Abeba
Certificates
About this item
ESD safety shoe made from smooth leather and a lightweight PU sole (fuel resistant).
- Perspiration-absorbent special lining with Silver Point
- Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- Lightweight PU sole with high slip resistance and shock-absorbing system
- Protective steel toecap (200 J)
- Foldable, adjustable heel strap
- Instep area with elastic band
Colour: White or black
Colour: White/black
Certifications: EN 61340 (ESD); DGUV 112-191; SB, A, E, FO, SRC.
Frequently Bought Together