Mailing boxes
Marketsource
Supplier: SARSTEDT
Jump to:
About this item
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
3 Options Available Below
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- SARS95.900Mailing boxes with integral restriants for 1 - 4 containersMailing boxes with integral restriants for 1 - 4 containers198×107×38 mmLog in for availability1 * 50 STSpecifications:MarketsourceMarketsource
- SARS95.901Mailing boxes with integral restriants for 1 - 4 containersMailing boxes with integral restriants for 1 - 4 containers198×107×50 mmLog in for availability1 * 50 ST35,85 €Specifications:MarketsourceMarketsource
- SARS95.902Mailing boxes with enclosed foam bag for 1 - 10 containersMailing boxes with enclosed foam bag for 1 - 10 containers220×170×40 mmLog in for availability1 * 50 ST31,29 €Specifications:MarketsourceMarketsource