Orders LinkContactUsLinkComponent
Micro tubes with thread
Micro tubes with thread
Micro tubes without skirted base, neutral cap, 2,0 ml, 1000/case
Catalog # SARS72.693.465
Marketsource
Supplier:   SARSTEDT
Micro tubes with thread
Catalog # SARS72.693.465
Supplier:  SARSTEDT
Marketsource
Micro tubes with thread
Micro tubes without skirted base, neutral cap, 2,0 ml, 1000/case
Price
177,10 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Specifications
  • Packed:
    1000/case
  • Capacity:
    2,0 ml
  • Description:
    Micro tubes without skirted base, neutral cap
  • Seq:
    02