Bosshead, cross type
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
About this item
Bosshead made of tempered cast iron, powder-coated.
- For rods up to 16 mm Ø
- With brass thumb or stainless steel safety heart screws
- Thread: M8
Weight: 180 g
2 Options Available Below
Product Details & Documents
Bosshead made of tempered cast iron, powder-coated.
- For rods up to 16 mm Ø
- With brass thumb or stainless steel safety heart screws
- Thread: M8
Weight: 180 g
Product Family Options
Product Information
- Spanning widthMaterialAvailabilityPrice
- NEUBM020-4850With brass thumb screws
- NEUBM020-4850SWith safety heart screws