Orders LinkContactUsLinkComponent
Cooling systems, CoolSafe™
Supplier:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Certificates
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™
20 Options Available Below
Product Details & Documents
Polystyrene cool safe box, aluminium cooling chamber.
  • Durable
  • Temperature stable
  • Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.
Product Family Options
CoolSafe Box with lid
Product Information
  • For
    No. of Holes
    Dimensions
    Availability
    Price
  • DIVECSF-BOX
    CoolSafe™ box, PS
    CoolSafe™ box, PS
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,80 €
    93,80 €
    1 * 1 ST
       93,80 €
Cooling chambers
Product Information
  • For
    No. of Holes
    Dimensions
    Availability
    Price
  • DIVECHAM-1000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    0,2 ml tubes
    96 in center (0,2 ml), 10 on side (4 for 1,5 ml tubes/6 for 0,5 ml tubes)
    156x79x25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 693,00 €
    693,00 €
    1 * 1 ST
       693,00 €
  • DIVECHAM-1020
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    0,2 ml PCR plates and tubes
    192
    156×171×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 320,00 €
    1 320,00 €
    1 * 1 ST
       1 320,00 €
  • DIVECHAM-2000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    0,5 ml tubes
    91
    156×79×32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 647,00 €
    647,00 €
    1 * 1 ST
       647,00 €
  • DIVECHAM-3000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    1,5 ml tubes
    48
    156×79×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 563,00 €
    563,00 €
    1 * 1 ST
       563,00 €
  • DIVECHAM-3050
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    1,5 ml cryogenic tubes
    55
    156×79×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 595,00 €
    595,00 €
    1 * 1 ST
       595,00 €
  • DIVECHAM-3840
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    384 well microplates
    384 in center, 10 on side (4 for 1,5 ml tubes/6 for o,5 ml tubes)
    156×79×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 080,00 €
    1 080,00 €
    1 * 1 ST
       1 080,00 €
  • DIVECHAM-4000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    5 ml ( 12×75 mm) tubes
    32
    156×79×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 693,00 €
    693,00 €
    1 * 1 ST
       693,00 €
  • DIVECHAM-5000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    0,2/0,5 and 1,5 ml tubes
    79 (20 for 1,5 ml/35 for 0,5 ml/24 for 0,2 ml)
    156×79×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 647,00 €
    647,00 €
    1 * 1 ST
       647,00 €
  • DIVECHAM-7050
    CoolSafe™ chambers, aluminium, 4 places
    CoolSafe™ chambers, aluminium, 4 places
    50 ml tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 612,00 €
    612,00 €
    1 * 1 ST
       612,00 €
  • DIVECHAM-8000
    CoolSafe™ chambers, aluminium
    CoolSafe™ chambers, aluminium
    1,5 ml tubes
    80
    210×67×32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 540,00 €
    540,00 €
    1 * 1 ST
       540,00 €
Cool bricks
Product Information
  • For
    No. of Holes
    Dimensions
    Availability
    Price
  • DIVEBRIK-2501
    Cool brick 1 °C
    Cool brick 1 °C
    0,2/0,5 and 1,5 ml tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,70 €
    96,70 €
    1 * 1 ST
       96,70 €
  • DIVEBRIK-1520
    Cool brick -10 °C
    Cool brick -10 °C
    0,2/0,5/1,5 and 5 ml tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
  • DIVEBRIK-1501
    Cool brick for 1 °C
    Cool brick for 1 °C
    5 ml tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,60 €
    99,60 €
    1 * 1 ST
       99,60 €
CoolSafe systems complete
Product Information
  • For
    No. of Holes
    Dimensions
    Availability
    Price
  • DIVECSAF-1000
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    0,2 ml PCR plates or tubes
    96 in center (0,2 ml), 10 on side (4 for 1,5 ml tubes/6 for 0,5 ml tubes)
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 999,00 €
    999,00 €
    1 * 1 ST
       999,00 €
  • DIVECSAF-2000
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    0,5 ml tubes
    91
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 970,00 €
    970,00 €
    1 * 1 ST
       970,00 €
  • DIVECSAF-3000
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    1,5 ml tubes
    48
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 843,00 €
    843,00 €
    1 * 1 ST
       843,00 €
  • DIVECSAF-3050
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    1,5 ml cryogenic tubes
    55
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 843,00 €
    843,00 €
    1 * 1 ST
       843,00 €
  • DIVECSAF-4000
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    5 ml (12×75 mm) tubes
    32
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 918,00 €
    918,00 €
    1 * 1 ST
       918,00 €
  • DIVECSAF-5000
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
    0,2/0,5 and 1,5 ml tubes
    79 (20 for 1,5 ml/35 for 0,5 ml/24 for 0,2 ml)
    140×216×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 866,00 €
    866,00 €
    1 * 1 ST
       866,00 €