Orders LinkContactUsLinkComponent
Multi-Safe sani 200 Secure Sharp Container
Multi-Safe sani 200 Secure Sharp Container
sani 200, 200 ml, 70x131 mm
Catalog # SARS77.3890.002
Marketsource
Supplier:   SARSTEDT
Multi-Safe sani 200 Secure Sharp Container
Catalog # SARS77.3890.002
Supplier:  SARSTEDT
Marketsource
Multi-Safe sani 200 Secure Sharp Container
Multi-Safe sani 200 Secure Sharp Container
sani 200, 200 ml, 70x131 mm
Price
175,16 €
Log in for availability
1 * 110 ST
Product Details & Documents
Boxes, made of PP, yellow/red. Secure sharp container with final closure in a handy size for doctors, nurses, rescue services- and ambulatory medical services, or for the doctors and rescue services supply bag.
  • Single-handed opening and closing of day lid
  • U-Shaped recess for sharps removal from Luer lock systems
Certifications: TRBA 250, UN, DIN EN ISO 23907
Specifications
  • Ø×H:
    70x131 mm
  • Capacity:
    200 ml
  • Description:
    sani 200