Orders LinkContactUsLinkComponent
Bowls, rectangular
Bowls, rectangular
Supplier:   Gilac PROFESSIONNEL
Bowls, rectangular
Certificates
Bowls, rectangular
6 Options Available Below
Product Details & Documents
LDPE, translucent.
  • Stackable
  • Durable
  • Available from 1 to 12 L
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 1 l
    160×160×65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,60 €
    6,60 €
    1 * 1 ST
       6,60 €
  • 3 l
    220×220×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,50 €
    11,50 €
    1 * 1 ST
       11,50 €
  • 5 l
    260×260×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,80 €
    14,80 €
    1 * 1 ST
       14,80 €
  • 7 l
    300×300×125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,60 €
    23,60 €
    1 * 1 ST
       23,60 €
  • 10 l
    350×350×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €
  • 12 l
    390×390×140 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 163,00 €
    163,00 €
    1 * 5 ST
       163,00 €