ContactUsLinkComponent
Spoon spatula, bent
Spoon spatula, bent
Catalog # USBE3317
undefined
Spoon spatula, bent
Catalog # USBE3317
Specifications
  • Length:
    185 mm
  • Environmentally Preferable:
  • Pk:
    1
  • Spoon size:
    19 mm
Specifications
About this item
Double-ended stainless steel spatula with bent spoon end and flat spatula end.