ContactUsLinkComponent
Adhesion slides for capillary gap stainers, HistoBond®
Adhesion slides for capillary gap stainers, HistoBond®
Catalog # MARI0890004
Supplier:  MARIENFELD
Adhesion slides for capillary gap stainers, HistoBond®
Catalog # MARI0890004
Supplier:  MARIENFELD
Specifications
  • Description:
    Adhesion slides for capillary gap stainers
  • Pk:
    2.500
Specifications
About this item
Adhesion slides are specially designed to endure harsh procedures.
  • Pre-cleaned and ready to use
  • Steps repeated until the slides tissue sections have been fully exposed to all required reagents
  • 80 μm high spacer stripe and a raised frosted end
Suitable for use with staining equipment in immunochemistry.
Certifications: ISO 8037/1
Packaging: Boxes of 50 pieces − 50 boxes per carton.