Order Entry
Belgium
ContactUsLinkComponent
Professional washing machine, Easy Wash, Basic Line, 60 cm, GW0160 series
Professional washing machine, Easy Wash, Basic Line, 60 cm, GW0160 series
Catalog # SMEGGW0160-1
Supplier:  SMEG
Professional washing machine, Easy Wash, Basic Line, 60 cm, GW0160 series
Catalog # SMEGGW0160-1
Supplier:  SMEG

Specifications

  • Capacity:
    150 L (washing chamber)
  • Temperature:
    85 °C
  • Description:
    Professional glassware washer, with pump for liquid neutraliser, stainless steel, basic line, GW0160-1
  • Seq:
    01
  • Model:
    GW0160-1
  • Chamber W×D×H:
    520×515×545 mm
  • Control:
    Microprocessor electronic control, electronic door lock system
  • Controller/programs:
    7 standard programs
  • Metering devices:
    Two liquid dispensers for powder reagents, mounted on the interior of the door
  • No. of washing levels:
    2
  • Power connection:
    230 V, (1 phase), 2,8 kW
  • Program operation/display:
    7 standard programs stored, LCD display
  • Water connection:
    Cold and demineralised water inlets
  • Weight:
    67 kg
  • W×D×H:
    600×605×850 mm
  • Pk:
    1

Specifications

About this item

Multipurpose washer for professional use able to wash up to 85 °C (thermal disinfection) and dose liquid detergent and neutraliser by peristaltic pumps. This washer represents a perfect choice to wash and disinfect all kinds of instruments used in small laboratories, universities, schools, specialised clinics, veterinary clinics and more.

  • Seven standard programs stored
  • Microprocessor electronic control
  • LCD with LED indicators for easy communication
  • Washing chamber and internal door in high grade AISI 304 stainless steel
  • Integrated water softener and salt sensor
  • Cold water and demineralised water connections
  • Electronic door lock system for user safety
  • Up two dosing peristaltic pumps for liquid dosing (liquid dosing improves the washing performance)
Delivery: Delivered without baskets, please contact your local Avantor sales office for details.