Storage Box, with Inserts
Supplier: NEOLAB
Jump to:
About this item
PS, clear.
- Insert tray weakly jams the tubes so it can be directly immersed into a water bath for quick- thawing the samples
- Base can be filled with dry ice or liquid nitrogen for storage, transport or snap-freezing
Outer dimensions: 185×110×65 mm
Inner dimensions: 165×90×55 mm
2 Options Available Below
Product Details & Documents
PS, clear.
- Insert tray weakly jams the tubes so it can be directly immersed into a water bath for quick- thawing the samples
- Base can be filled with dry ice or liquid nitrogen for storage, transport or snap-freezing
Outer dimensions: 185×110×65 mm
Inner dimensions: 165×90×55 mm
Product Family Options
Product Information
- MaterialAvailabilityPrice
- NEOL4-5011for 0,5 ml tubes, 6×12 compartments
- NEOL4-5010for 1,5 ml tubes, 5×10 compartments
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



