Order Entry
Belgium
Orders LinkContactUsLinkComponent
Safety baskets, PP

Safety baskets, PP

Rectangular basket with handle, blue, 28 l, 380×230×255 mm
Catalog # NEOL1-7011
Supplier:   NEOLAB
Safety baskets, PP
Catalog # NEOL1-7011
Supplier:  NEOLAB
Safety baskets, PP
Safety baskets, PP

Rectangular basket with handle, blue, 28 l, 380×230×255 mm

Price
15,91 €
Log in for availability
1 * 1 ST
Product Details & Documents

Laboratory basket made of PP with foldable handle.

  • Good chemical resistance
  • Good temperature resistance

Stackable, also usable as a container for immersing glassware in cleaning solutions or for drying. The black handles are made from recycled material.

Specifications
  • Capacity:
    28 l
  • Description:
    Rectangular basket with handle, blue
  • W×D×H:
    380×230×255 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.