Attachable screwthread tubes
GL 18
Catalog # SMIT746/GL18
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread GL.
Specifications
- Thread:GL 18
- Seq:02