Orders LinkContactUsLinkComponent
Sample loops for Valco valves
Sample loops for Valco valves
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Supplier:  VICI JOUR RESEARCH
Certificates
Sample loops for Valco valves
8 Options Available Below
Product Details & Documents
These VICI Jour sample loops are made of high quality stainless steel 316.
  • Fits all Valco W and UW type valves
  • High pressure resistance, low permeability
  • Tolerance of ±0,05 mm and pressure rating of <350 bar
They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
Delivery information: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.
Product Family Options
Product Information
  • Material
    Capacity
    Availability
    Price
  • JOURJR-SLV-76-5
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    5 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €
  • JOURJR-SLV-76-10
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    10 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €
  • JOURJR-SLV-76-20
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    20 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €
  • JOURJR-SLV-76-50
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    50 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €
  • JOURJR-SLV-76-100
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    100 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €
  • JOURJR-SLV-76-250
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    250 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,80 €
    83,80 €
    1 * 1 ST
       83,80 €
  • JOURJR-SLV-76-500
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    500 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,50 €
    91,50 €
    1 * 1 ST
       91,50 €
  • JOURJR-SLV-76-1000
    Sample loop, for Valco W and UW type valves
    Sample loop, for Valco W and UW type valves
    Stainless steel
    1000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €