ContactUsLinkComponent
Rotary valves with cap connection
Rotary valves with cap connection
Catalog # OMNI001113
Supplier:  OMNIFIT
undefined
Rotary valves with cap connection
Catalog # OMNI001113
Supplier:  OMNIFIT
Specifications
  • Description:
    Valve, 4-port, connects 3 ports in a T
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Valves are constructed from PTFE with a PCTFE rotor and have a 1,5 mm bore size. They accept tubing sizes between 0,5 and 4 mm Ext.Ø. and are pressure rated to 50 psi.