ContactUsLinkComponent
Gel loading pipette tips, MµltiFlex®
Gel loading pipette tips, MµltiFlex®
Catalog # 612-0675
Supplier:  Roth Carl
Gel loading pipette tips, MµltiFlex®
Catalog # 612-0675
Supplier:  Roth Carl
Specifications
  • Packed:
    200/rack
  • Sterile:
    -
  • Volume:
    0,5 - 200 µl
  • Environmentally Preferable:
  • Tip style:
    Flat, 0,2 mm
  • Tip colour:
    Transparent
  • Pk:
    200
Specifications
About this item
These tips are suitable for loading SDS page protein gels, horizontal agarose and DNA sequencing gels.
  • Free of RNAse/DNAse and pyrogens
  • Free of human DNA and PCR inhibitors
  • Autoclavable
Suitable for Brand, Eppendorf, Finnpipette®, Gilson, Oxford, Roth, Sartorius, Socorex.