Digital refractometers
0 − 45 °Bx, ±0,001 °Bx
Catalog # 635-0068
Supplier: EUROMEX
Jump to:
Product Details & Documents
Digital refractometer with automatic temperature compensation (ATC) for sugar concentration.
Delivery information: Supplied with a nine-volt battery.
Specifications
- Range:0 − 45 °Bx
- Precision:±0,001 °Bx