Orders LinkContactUsLinkComponent
Sample loops for Valco valves
Sample loops for Valco valves
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Supplier:  VICI JOUR RESEARCH
Certificates
Sample loops for Valco valves
8 Options Available Below
Product Details & Documents
These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.
  • Fits Valco W and UW type valves
  • Tolerance of ±0,025 mm
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.
Delivery information: Supplied with PEEK hex-head nuts and PEEK ferrules.
Product Family Options
Product Information
  • Material
    Capacity
    Pressure
    Availability
    Price
  • Sample loop
    PEEK
    5 µl
    max. 400 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,00 €
    72,00 €
    1 * 1 ST
       72,00 €
  • Sample loop
    PEEK
    10 µl
    max. 400 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,00 €
    72,00 €
    1 * 1 ST
       72,00 €
  • Sample loop
    PEEK
    20 µl
    max. 400 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,30 €
    75,30 €
    1 * 1 ST
       75,30 €
  • Sample loop
    PEEK
    50 µl
    max. 400 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,30 €
    75,30 €
    1 * 1 ST
       75,30 €
  • Sample loop
    PEEK
    100 µl
    max. 400 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,80 €
    77,80 €
    1 * 1 ST
       77,80 €
  • Sample loop
    PEEK
    250 µl
    max. 240 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • Sample loop
    PEEK
    500 µl
    max. 240 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • Sample loop
    PEEK
    1000 µl
    max. 240 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €