ContactUsLinkComponent
Counting chamber, Modified Fuchs-Rosenthal
Counting chamber, Modified Fuchs-Rosenthal
Catalog # HAWKAC6000
Supplier:  HAWKSLEY AND SONS
Counting chamber, Modified Fuchs-Rosenthal
Catalog # HAWKAC6000
Supplier:  HAWKSLEY AND SONS
Specifications
  • Depth of chamber:
    0,2 mm
  • Type:
    Fuchs-Rosenthal
  • Description:
    Counting chamber
  • Pk:
    1
Specifications
About this item
This two cell counting chamber has metallised platforms.
  • Total area of ruling: 1/6 mm²
  • Rhodium coating
Delivery: Supplied with two cover glasses.