Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Oasis® HLB
Solid phase extraction cartridges, Oasis® HLB
Marketsource
Supplier:   WATERS
Solid phase extraction cartridges, Oasis® HLB
Supplier:  WATERS
Marketsource
Certificates
Solid phase extraction cartridges, Oasis® HLB
3 Options Available Below
Product Details & Documents
Oasis® HLB cartridges contain the Oasis® HLB sorbent, which is a universal polymeric reversed phase sorbent that was developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
Since the HLB sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading.
These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Product Family Options
Product Information
  • Column Format
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • WATEWAT106202
    Oasis® HLB 6 cc Vac cartridge
    Oasis® HLB 6 cc Vac cartridge
    Cartridge
    80 Å
    30 µm
    6 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 214,00 €
    214,00 €
    1 * 30 ST
       214,00 €
    Specifications:
    Marketsource
    Marketsource
  • WATE186000115
    Oasis® HLB 6 cc Vac cartridge
    Oasis® HLB 6 cc Vac cartridge
    Cartridge
    80 Å
    60 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 308,00 €
    308,00 €
    1 * 30 ST
       308,00 €
    Specifications:
    Marketsource
    Marketsource
  • WATE186000117
    Oasis® HLB 20 cc Vac cartridge
    Oasis® HLB 20 cc Vac cartridge
    Cartridge
    80 Å
    60 µm
    20 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 403,00 €
    403,00 €
    1 * 20 ST
       403,00 €
    Specifications:
    Marketsource
    Marketsource