ContactUsLinkComponent
Microcentrifuge tubes
Microcentrifuge tubes
Catalog # DELT200400P
Supplier:  DELTALAB
Microcentrifuge tubes
Catalog # DELT200400P
Supplier:  DELTALAB
Specifications
  • Type:
    with attached lid, Eppendorf type
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Pk:
    6.000
Specifications
About this item
Manufactured in polypropylene.
  • Autoclavable
  • Easy opening and closing
  • Graduated tube
  • It features a frosted area for writing
  • Flat cap
  • Will withstand centrifugation up to: 12500×g