Order Entry
Belgium
ContactUsLinkComponent
Test tube racks, Combi
Test tube racks, Combi
Catalog # 212-9175
Supplier:  DELTALAB
Test tube racks, Combi
Catalog # 212-9175
Supplier:  DELTALAB

Specifications

  • For tubes:
    0,5/1,5/2,0/15/50 ml
  • No. of Holes:
    12/9/6/2
  • W×D×H:
    48×92×92 mm
  • Pk:
    1

Specifications

About this item

Made of polypropylene, this combi rack can be autoclaved. This design consists of a unit of four independent cubes packaged in a clear support tray.

  • Excellent chemical resistance
  • Excellent temperature resistance

Each rack may be independently rotated to allow the simultaneous storage and use of multiple sizes of tubes (micro tubes to 50 ml tubes). Each side of the racks is alpha-numeric matrixed for better sample location.

The following tubes can be accommodated in each rack:
- 2×50 ml conical tubes,
- 6×15 ml conical tubes,
- 9×12 mm diameter tubes or 1,5/2,0 ml micro tubes
- 12×0,5 ml micro tubes.