Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Macro cuvette

Macro, 340 – 900 nm, 45 mm, 4 ml, PS
  634-8211
Macro cuvette
  634-8211
Macro cuvette

Macro, 340 – 900 nm, 45 mm, 4 ml, PS

 
159,00 €
   
1 * 1.000 ST
 

Designed for enzymatic determinations, since the very thin walls enable rapid and uniform tempering.

  • Manufactured from crystal clear polystyrene
  • Characterised by minimal light scatter
  • High transmission
 
  • Type:
    Macro
  • Volume:
    4 ml
  • Window material:
    PS
  • Height:
    45 mm
  • Range:
    340 – 900 nm