Sample storage convenience kits
Supplier: THERMO NATIONAL SCIENTIFIC
Aller à :
À propos de cet article
These sample storage unassembled convenience kits save time during sample preparation.
- Solid top convenience kits include matched quantities of vials and screw caps with pre-assembled septa
- Open top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags
4 Options disponibles ci-dessous
Product Details & Documents
These sample storage unassembled convenience kits save time during sample preparation.
- Solid top convenience kits include matched quantities of vials and screw caps with pre-assembled septa
- Open top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags
Documents
Product Family Options
Convenience kit, clear
Informations sur le produit
- CapacityØ×HCap includedCap typeCap materialCap colourSeptumThicknessAvailabilityPrice
- NSCAB7990-2Convenience kitsConvenience kits4,00 ml15×45 mmYesOpen topPPBlackWhite PTFE/clear silicone0,01/0,05"Connexion Pour la disponibilité1 * 100 ST71,50 €
- NSCAB7990-3Convenience kitsConvenience kits8,00 ml17×60 mmYesOpen topPPBlackWhite PTFE/clear silicone0,01/0,05"Connexion Pour la disponibilité1 * 200 ST167,00 €
- NSCAB7990-12Convenience kitsConvenience kits12,00 ml19×65 mmYesOpen topPPBlackWhite PTFE/clear silicone0,01/0,05"Connexion Pour la disponibilité1 * 200 ST182,00 €
- NSCAB7990-4Convenience kitsConvenience kits16,00 ml21×70 mmYesOpen topPPBlackWhite PTFE/clear silicone0,01/0,05"Connexion Pour la disponibilité1 * 200 ST221,00 €