Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
Certificates
Flasks, flat bottom with standard ground joint
5 Options Available Below
Product Details & Documents
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 50 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,50 €
    40,50 €
    1 * 1 ST
       40,50 €
  • 100 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,60 €
    42,60 €
    1 * 1 ST
       42,60 €
  • 250 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
  • 100 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,60 €
    42,60 €
    1 * 1 ST
       42,60 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €