Order Entry
Belgium
ContactUsLinkComponent
Tablet bottles, with PE cap
Tablet bottles, with PE cap
  215-2011
undefined
Tablet bottles, with PE cap
  215-2011

 

  • Ext.Ø:
    12,75 mm
  • Height:
    40 mm
  • Seq:
    01
  • Pk:
    200

 

 

Clear glass.

  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust

With plug-in stopper.