ContactUsLinkComponent
Bottles, without pump vaporiser, BOSTON
Bottles, without pump vaporiser, BOSTON
Catalog # MUEL14.289.20
Supplier:  MUELLER KREMPEL
undefined
Bottles, without pump vaporiser, BOSTON
Catalog # MUEL14.289.20
Supplier:  MUELLER KREMPEL
Specifications
  • Type:
    BOSTON type, with GL 20 thread
  • Capacity:
    100 ml
  • Description:
    Cosmetic bottles
  • Pk:
    20
Specifications
About this item
Cosmetic bottles, made of PET, clear.
  • Good chemical resistance
  • Good temperature resistance
  • Pump vaporiser to be ordered separately
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904