pH electrodes
Supplier: KNICK
Jump to:
About this item
3 Options Available Below
Product Details & Documents
Product Family Options
Electrodes
Product Information
- TypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnectionForAvailabilityPrice
- 662-3531SE 103 N, pH electrodeSE 103 N, pH electrodeSE 103 N0...14−5...+100 °CGlass3 mol/l KClCeramic12×170 mmDINLaboratory purposes, industrial production, processing systemsLog in for availability1 * 1 ST281,00 €
- 662-3535SE 104 N, pH puncture electrode, pH 2 -13SE 104 N, pH puncture electrode, pH 2 -13SE 104 N2...13−5...+80 °CPlasticPolymerHole junction12×99 mmDINSemi-solid substancesLog in for availability1 * 1 ST425,00 €
- KNICSE106NSE 106 N, combination pH/PT1000 electrodeSE 106 N, combination pH/PT1000 electrodeSE 106 N0...14−5...+100 °CGlass3 mol/l KClGround glass sleeve12×170 mmDINDifficult samples, low conductivity samples, coating mediaLog in for availability1 * 1 ST496,00 €
Electrodes
Product Information
- Catalog #ConnectionTemp.Diaphr.Ø×LForMaterialpHDescriptionTypeElectrolyteAvailabilityPrice
- DIN coax connector and 4 mm spring connector−5...+100 °CCeramic12×110 mmPrecision measurements, chemical applications, aggressive media, quality controlGlass0...14pH sensor with integrated temperature detectorSE 102 ANLiquid, 3 mol/L KClLog in for availability1 * 1 ST0,00 €