Orders LinkContactUsLinkComponent
Soxhlet extractor, multiposition
Soxhlet extractor, multiposition
Soxhlet extraction, 100 ml, 6 sample pieces, SMX 100, 100 ml
Catalog # BEHRB00218423
Supplier:   BEHR
Soxhlet extractor, multiposition
Catalog # BEHRB00218423
Supplier:  BEHR
Soxhlet extractor, multiposition
Soxhlet extraction, 100 ml, 6 sample pieces, SMX 100, 100 ml
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Complete workstation to undertake Soxhlet extraction using 6×100 ml flasks.
Delivery information: Workstation comprises temperature and time controller, heating block with six sample positions, condenser frame with coolant water distribution manifold, glassware including flasks, extractors and Dimroth condensers.
Specifications
  • Type:
    SMX 100
  • Description:
    Soxhlet extraction, 100 ml, 6 sample pieces
  • Flask capacity:
    100 ml