ContactUsLinkComponent
Cell culture media, Schneider's Insect Medium
Cell culture media, Schneider's Insect Medium
Catalog # L0207-500
Supplier:  Biowest
CAS Number:  
undefined
Cell culture media, Schneider's Insect Medium
Catalog # L0207-500
Supplier:  Biowest
CAS Number:  
Specifications
  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    Schneider's Insect Medium
Specifications
About this item
Schneider’s Insect Medium was developed to support the growth of excised imaginal discs from the fruit fly, Drosophila melanogaster. When supplemented with 5 to 20% heat-inactivated foetal bovine serum Schneider's mediuam has been found to support the rapid growth of both primary and established cultures of cells derived from Drosophila spp. and several other dipterans.