MS interface restrictors, QuickSwap
Supplier: Agilent
Certificates
About this item
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Certified headspace crimp top vials
Supplier: Agilent
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Avantor® ACE® C8, HPLC Columns, 3 µm
Supplier: Avantor
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Modified syringes Microliter™7000 series
Supplier: HAMILTON BONADUZ
Porapak T 80 - 100 mesh, 31 g 1 * 1 items
Supplier: WATERS
Snap top solvent vials and closures, 19 mm, CrossLab
Supplier: Agilent
Caps for LC vials
Supplier: Agilent
TLC/HPTLC plates cutter, smartCut
Supplier: CAMAG
VWR®, Aluminium Crimp Caps, ND11 with Central Hole, 5,5 mm and Septum
Supplier: LA PHA PACK
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Screw caps and septa for storage vials, Chromacol™
Supplier: CHROMACOL
VWR® ACE® UltraCore Super C18 HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
CHROMABOND QUECHERS MIX I STICKS EXTRACT 1 * 50 items
Supplier: MACHEREY-NAGEL
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
Reusable half masks, Advantage 200 LS
Supplier: M S A
NEEDLE 1260 INFINITY AUTOSAMPLER 1 * 1 items
Supplier: Agilent
Gas-clean filters
Supplier: VARIAN