Orders LinkContactUsLinkComponent
Test fluids for viscometers
Test fluids for viscometers
Supplier:   HAAKE
Test fluids for viscometers
Supplier:  HAAKE
Certificates
Test fluids for viscometers
7 Options Available Below
Product Details & Documents
Test and certificated fluids for viscometery - mesurement temperature at 20 °C unless indicated.
Caution: * Measurement temperature 25 °C
Documents
Product Family Options
Product Information
  • Type
    Viscosity
    Pk
    Availability
    Price
  • E7
    5
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 209,00 €
    209,00 €
    1 * 100 mL
       209,00 €
  • E200
    120
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 209,00 €
    209,00 €
    1 * 100 mL
       209,00 €
  • E2000
    1900
    100 mL
    Discontinued
  • E6000
    6000
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 209,00 €
    209,00 €
    1 * 100 mL
       209,00 €
  • E15000
    15000
    100 mL
    Discontinued
  • S30000* (silicone oil)
    ~30000
    500 mL
    Discontinued
  • E40000
    40000
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 203,00 €
    203,00 €
    1 * 100 mL
       203,00 €
  • S100* (silicone oil)
    ~100
    500 mL
    Discontinued
  • S1000* (silicone oil)
    ~1000
    500 mL
    Discontinued
  • E1300*
    ~1300
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 438,00 €
    438,00 €
    1 * 500 mL
       438,00 €
  • E33*
    ~33
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 438,00 €
    438,00 €
    1 * 500 mL
       438,00 €
  • E330*
    ~330
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 438,00 €
    438,00 €
    1 * 500 mL
       438,00 €