ContactUsLinkComponent
VWR® PolarSafe, Dispenser
VWR® PolarSafe, Dispenser
Catalog # 525-0672
Supplier:  VWR Collection
undefined
VWR® PolarSafe, Dispenser
Catalog # 525-0672
Supplier:  VWR Collection
Specifications
  • Colour:
    Blue
  • Description:
    Acryl rack for cryo labels and dots
  • Pk:
    1
Specifications
About this item
Addition to the PolarSafe family of products, the label dispenser can accommodate up to six rolls of cryogenic dots or strip labels. Made from durable, easy-to-clean, acrylic plastic, the dispenser has been designed to conveniently dispense labels for your cryogenic vials, tubes or plates.
  • Four rubber feet hold item in place when used on countertops
  • Can also be wall mounted (screws not included)